Merck
  • Improvement of the solubilization of proteins in two-dimensional electrophoresis with immobilized pH gradients.
Improvement of the solubilization of proteins in two-dimensional electrophoresis with immobilized pH gradients.
Electrophoresis (1997-03-01)
T Rabilloud, C Adessi, A Giraudel, J Lunardi
ABSTRACT
Membrane and nuclear proteins of poor solubility have been separated by high resolution two-dimensional (2-D) gel electrophoresis. Isoelectric focusing with immobilized pH gradients leads to severe quantitative losses of proteins in the resulting 2-D map, although the resolution is usually high. Protein solubility could be improved by using denaturing solutions containing various detergents and chaotropes. Best results were obtained with a denaturing solution containing urea, thiourea, and detergents (both nonionic and zwitterionic). The usefulness of thiourea-containing denaturing mixtures is shown for microsomal and nuclear proteins as well as for tubulin, a protein highly prone to aggregation.
Sigma-Aldrich
3-(Benzyldimethylammonio)propanesulfonate, BioXtra, ≥99.0% (HPCE)