Merck
  • Seleno compounds and glutathione peroxidase catalyzed decomposition of S-nitrosothiols.
Seleno compounds and glutathione peroxidase catalyzed decomposition of S-nitrosothiols.
Biochemical and biophysical research communications (1996-11-01)
Y Hou, Z Guo, J Li, P G Wang
ABSTRACT
Seleno compounds such as selenocystamine and seleno-D, L-cystine were found to catalyze the decomposition of S-nitrosothiols (e.g. S-nitroso-glutathione and S-nitroso-N-acetyl-D, L-penicillamine) in the presence of different thiols (e.g. glutathione, N-acetyl-D-penicillamine and 2-mercaptoethanol), and liberate nitric oxide. It was also found that glutathione peroxidase itself can catalyze the decomposition of S-nitrosoglutathione without the presence of any thiol or H2O2.
Sigma-Aldrich
Selenocystamine dihydrochloride, powder
Pricing and availability is not currently available.