- Status asthmaticus in a paramedic following exposure to a roadside flare: a case report.
Status asthmaticus in a paramedic following exposure to a roadside flare: a case report.
The Journal of emergency medicine (1997-01-01)
J H Federman, J J Sachter
PMID9017492
ABSTRACT
Highway flares are commonly used by police and emergency service workers to divert traffic away from roadside accidents. We report a case of a 35-yr-old paramedic who developed a sudden, severe asthmatic reaction following a brief exposure to such a flare, necessitating intubation and a prolonged hospitalization.