Merck
  • Individual and combined effects of fumonisin B1 present in Fusarium moniliforme culture material and diacetoxyscirpenol or ochratoxin A in turkey poults.
Individual and combined effects of fumonisin B1 present in Fusarium moniliforme culture material and diacetoxyscirpenol or ochratoxin A in turkey poults.
Poultry science (1997-02-01)
L F Kubena, T S Edrington, R B Harvey, T D Phillips, A B Sarr, G E Rottinghaus
ABSTRACT
The individual and combined effects of feeding diets containing 300 mg fumonisin B1 (FB1), and 4 mg diacetoxyscirpenol (DAS) or 3 mg ochratoxin A (OA) were evaluated in two experiments using female turkey poults (Nicholas Large Whites) from day of hatch to 3 wk of age. When compared with controls, body weight gains were reduced 30% (Study 1) and 24% (Study 2) by FB1, 30% by DAS, 8% by OA, 46% by the FB1 and DAS combination, and 37% by the FB1 and OA combination. The efficiency of feed utilization was adversely affected by all treatments except FB1 in Experiment 2. Relative weights of the liver were significantly increased by all treatments except the DAS treatment. Serum concentrations of cholesterol were decreased and activities of aspartate aminotransferase and lactate dehydrogenase were increased and several hematological values were altered in poults fed FB1 alone and in combination with either DAS or OA. Results indicate additive or less than additive toxicity, but not toxic synergy, when poults are fed diets containing 300 mg FB1, and 4 mg DAS or 3 mg OA/kg of diet. The likelihood of encountering FB1, DAS, or OA at these concentrations in finished feed is small. However, under field conditions, other stress factors could alter the impact of these mycotoxins on the health and performance of poultry.
Product Description
Supelco
Diacetoxyscirpenol solution, ~100 μg/mL in acetonitrile, analytical standard