Merck
  • A novel phospholipase A2 activity in saliva of the lone star tick, Amblyomma americanum (L.).
A novel phospholipase A2 activity in saliva of the lone star tick, Amblyomma americanum (L.).
Experimental parasitology (1997-10-27)
A S Bowman, C L Gengler, M R Surdick, K Zhu, R C Essenberg, J R Sauer, J W Dillwith
ABSTRACT
Saliva from female lone star ticks, Amblyomma americanum, contained a novel phospholipase A2 (PLA2) activity that hydrolyzed 14C-arachidonate from 14C-arachidonyl phosphatidylcholine. The tick saliva PLA2 (ts-PLA2) was active over a broad pH range (4.5-11.5) with two distinct pH optima of pH 5.5 and 9.5. Though extracellular PLA2s are reported to be activated by millimolar Ca2+, ts-PLA2 was sensitive to submicromolar Ca2+ and was half-maximally activated by 3.5 microM Ca2+. Tick saliva contains > 500 microM Ca2+ and the feeding lesion in the host is expected to contain millimolar Ca2+. Saliva exhibited a single peak of PLA2 activity corresponding to a molecular weight of 55.7 +/- 1.3 kDa by size exclusion chromatography. The ts-PLA2 was unaffected by a variety of compounds known to inhibit either secreted or cytosolic PLA2s from other sources. However, ts-PLA2 was inhibited by the substrate analog, oleyloxyethyl phosphorylcholine (IC50 = 1.4 microM), and the end product, arachidonic acid (IC50 = 38 microM). Low concentrations of dithiothreitol did not greatly affect ts-PLA2, but activity was reduced at higher concentrations. The PLA2 activity found in A. americanum salivary glands showed many similarities to ts-PLA2, but also some distinct differences. Secreted at the tick-host interface, ts-PLA2 is thought to play an important, but unknown, role during the prolonged tick feeding.