Merck
  • Ascorbate-induced oxidative inactivation of Zn2+-glycerophosphocholine cholinephosphodiesterase.
Ascorbate-induced oxidative inactivation of Zn2+-glycerophosphocholine cholinephosphodiesterase.
Journal of neurochemistry (1998-03-07)
D E Sok
ABSTRACT
Zn2+-glycerophosphocholine cholinephosphodiesterase, responsible for the conversion of glycerophosphocholine into glycerol and phosphocholine, was inactivated during incubation with ascorbic acid at 38 degrees C. The inclusion of copper ions or Fe2+ accelerated the ascorbate-induced inactivation, with Cu2+ or Cu+ being much more effective than Fe2+, suggestive of ascorbate-mediated oxidation. Dehydroascorbic acid had no effect on the phosphodiesterase, but H2O2 inactivated the enzyme in a concentration-dependent manner. Also, the enzyme was inactivated partially by a superoxide anion-generating system but not an HOCl generator. In support of involvement of H2O2 in the ascorbate action, catalase and superoxide dismutase expressed a complete and a partial protection, respectively. However, hydroxy radical scavengers such as mannitol, benzoate, or dimethyl sulfoxide were incapable of preventing the ascorbate action, excluding the participation of extraneous .OH. Although p-nitrophenylphosphocholine exhibited a modest protection against the ascorbate action, a remarkable protection was expressed by amino acids, especially by histidine. In addition, imidazole, an electron donor, showed a partial protection. Separately, when Cu2+-induced inactivation of the phosphodiesterase was compared with the ascorbate-mediated one, the protection and pH studies indicate that the mechanism for the ascorbate action is different from that for the Cu2+ action. Here, it is proposed that Zn2+-glycerophosphocholine cholinephosphodiesterase is one of brain membrane proteins susceptible to oxidative inactivation.
Sigma-Aldrich
Xanthine Oxidase microbial, lyophilized powder, ≥7 units/mg solid
Sigma-Aldrich
Xanthine Oxidase from bovine milk, Grade I, ammonium sulfate suspension, ≥0.4 units/mg protein
Sigma-Aldrich
Xanthine Oxidase from bovine milk, Grade IV, ammonium sulfate suspension, ≥0.1 units/mg protein
Sigma-Aldrich
Xanthine Oxidase from bovine milk, lyophilized powder, 0.4-1.0 units/mg protein
Sigma-Aldrich
Xanthine Oxidase from bovine milk, Grade III, ammonium sulfate suspension, ≥0.8 units/mg protein