Merck
Cyanoglycosylation products of 17-O-acetyl-testosterone.
Carbohydrate research (1998-11-20)
A Tobari, T Shimizu, H Miyamae, A Nagasawa, M Kawase, S Saito
ABSTRACT
17-O-Acetyl testosterone, which has no susceptible hydroxyl or carboxyl group for glycosylation, was glycosylated with 2,3,4,6-tetra-O-acetyl-alpha-D-glucopyranosyl bromide in the presence of a mixed catalyst, Hg(CN)2 and HgBr2, in benzene-nitromethane. Reaction occurred on the alpha, beta-unsaturated ketone on the six-membered A-ring to give six 3-O-glycosides, each bearing a cyano group at the 3- or 5-position of the aglycon, and a 3-O-glycoside bearing a CONH2 group at the 3-position. Structural analyses of these products were carried out by various NMR (1H, 13C NMR, 1H-1H and 1H-13C COSY, HMBC, and DEPT), FABMS and X-ray analyses. The mechanisms of the formations of the products are discussed. It was determined that mercuric cyanide was essential as a catalyst for the progress of the cyanoglycosylation.
Sigma-Aldrich
Mercury(II) bromide, ACS reagent
Sigma-Aldrich
Mercury(II) bromide, puriss. p.a., ACS reagent, ≥99.0% (precipitation titration)