Merck
  • Liquid chromatographic detection of Dowtherm A contamination in oleochemicals and edible oils.
Liquid chromatographic detection of Dowtherm A contamination in oleochemicals and edible oils.
Journal of AOAC International (1999-08-13)
F M Moh, T S Tang
ABSTRACT
A rapid and direct liquid chromatographic (LC) technique is described for the determination of a eutectic mixture of diphenyl oxide and biphenyl such as Dowtherm A thermal heating fluid (THF) in oleochemicals and palm olein. Analysis is performed with an RP-18 column with fluorescence detection (excitation at 247 nm and emission at 310 nm). The isocratic mobile phase (1.0 mL/min) consists of methanol and water (90 + 10, v/v). A linear calibration model (correlation coefficient = 0.9999) was developed directly from used Dowtherm A THF with the biphenyl peak (4.70 min) as a marker. Average recoveries from spiked glycerin, fatty alcohol mixture, methyl ester mixture, fatty acids, and palm olein were 90.9-108.7%, with a detection limit of 0.1 microgram/mL. The technique requires no prior sample cleanup nor extraction steps and is good for quality assurance purposes.
Sigma-Aldrich
Dowtherm® A, eutectic mixture of 26.5% diphenyl + 73.5% diphenyl oxide