Merck
New experiments on HNCSe and HCNSe radical cations.
Rapid communications in mass spectrometry : RCM (2005-12-07)
P Gerbaux, N Dechamps, R Flammang, P Nagi Reddy, R Srinivas
ABSTRACT
Dissociative ionization of the selenourea Se=C(NH(2))(2) (2) conveniently generates beams of pure isocyanoselenic acid radical cations. The HNCSe(.+) connectivity is established by collisional activation and by associative ion-molecule reactions with dimethyl sulfide or nitric oxide using a large-scale hybrid mass spectrometer.
Sigma-Aldrich
Selenourea, 98%