Merck
  • Nucleotide sequence of the iap gene, responsible for alkaline phosphatase isozyme conversion in Escherichia coli, and identification of the gene product.
Nucleotide sequence of the iap gene, responsible for alkaline phosphatase isozyme conversion in Escherichia coli, and identification of the gene product.
Journal of bacteriology (1987-12-01)
Y Ishino, H Shinagawa, K Makino, M Amemura, A Nakata
ABSTRACT
The iap gene in Escherichia coli is responsible for the isozyme conversion of alkaline phosphatase. We analyzed the 1,664-nucleotide sequence of a chromosomal DNA segment that contained the iap gene and its flanking regions. The predicted iap product contained 345 amino acids with an estimated molecular weight of 37,919. The 24-amino-acid sequence at the amino terminus showed features characteristic of a signal peptide. Two proteins of different sizes were identified by the maxicell method, one corresponding to the Iap protein and the other corresponding to the processed product without the signal peptide. Neither the isozyme-converting activity nor labeled Iap proteins were detected in the osmotic-shock fluid of cells carrying a multicopy iap plasmid. The Iap protein seems to be associated with the membrane.
Sigma-Aldrich
CRISPR/Cas9 Products and Services, Design and order CRISPR gRNA, Cas9, screening libraries, controls and companion products. Formats include plant, lentivirus, IVT-RNA, plasmid, synthetic, and protein.