Merck
  • The hallucinogen N,N-dimethyltryptamine (DMT) is an endogenous sigma-1 receptor regulator.
The hallucinogen N,N-dimethyltryptamine (DMT) is an endogenous sigma-1 receptor regulator.
Science (New York, N.Y.) (2009-02-14)
Dominique Fontanilla, Molly Johannessen, Abdol R Hajipour, Nicholas V Cozzi, Meyer B Jackson, Arnold E Ruoho
ABSTRACT
The sigma-1 receptor is widely distributed in the central nervous system and periphery. Originally mischaracterized as an opioid receptor, the sigma-1 receptor binds a vast number of synthetic compounds but does not bind opioid peptides; it is currently considered an orphan receptor. The sigma-1 receptor pharmacophore includes an alkylamine core, also found in the endogenous compound N,N-dimethyltryptamine (DMT). DMT acts as a hallucinogen, but its receptor target has been unclear. DMT bound to sigma-1 receptors and inhibited voltage-gated sodium ion (Na+) channels in both native cardiac myocytes and heterologous cells that express sigma-1 receptors. DMT induced hypermobility in wild-type mice but not in sigma-1 receptor knockout mice. These biochemical, physiological, and behavioral experiments indicate that DMT is an endogenous agonist for the sigma-1 receptor.
Sigma-Aldrich
N-ω-Methyltryptamine, 99%
Pricing and availability is not currently available.