Merck
  • Determination of glycarbylamide in chicken tissue by high-performance liquid chromatography.
Determination of glycarbylamide in chicken tissue by high-performance liquid chromatography.
Journal of AOAC International (1999-04-07)
Y Yamamoto, F Kondo
ABSTRACT
A high-performance liquid chromatographic (HPLC) method for determining glycarbylamide (GB) in chicken tissue was developed. GB was extracted with acetonitrile, followed by solid-phase extraction cleanup using a Bond Elut cartridge column with neutral alumina. After the extract had been evaporated to dryness, the residue was dissolved in 1.0 mL 0.1 N sodium hydroxide. Then 1.0 mL 0.1 M potassium dihydrogen phosphate solution was added to it. HPLC separation was done on a 250 x 4.6 mm id TSK-GEL ODS 80 column with 0.05M potassium dihydrogen phosphate as the mobile phase. Ultraviolet detection was done at a wavelength of 260 nm. The calibration curve of standard GB solutions was linear between 0.16 and 3 micrograms/mL (correlation coefficient, r = 0.999). The recovery of GB from chicken muscle spiked at 0.8 microgram/g was 88.6 +/- 2.3% (mean +/- standard deviation, n = 5), and the lower limit of determination was 0.05 microgram/g in chicken muscle.