Merck
  • Ligand binding by antibody IgE Lb4: assessment of binding site preferences using microcalorimetry, docking, and free energy simulations.
Ligand binding by antibody IgE Lb4: assessment of binding site preferences using microcalorimetry, docking, and free energy simulations.
Biophysical journal (1999-06-04)
C A Sotriffer, W Flader, A Cooper, B M Rode, D S Linthicum, K R Liedl, J M Varga
ABSTRACT
Antibody IgE Lb4 interacts favorably with a large number of different compounds. To improve the current understanding of the structural basis of this vast cross-reactivity, the binding of three dinitrophenyl (DNP) amino acids (DNP-alanine, DNP-glycine, and DNP-serine) is investigated in detail by means of docking and molecular dynamics free energy simulations. Experimental binding energies obtained by isothermal titration microcalorimetry are used to judge the results of the computational studies. For all three ligands, the docking procedure proposes two plausible subsites within the binding region formed by the antibody CDR loops. By subsequent molecular dynamics simulations and calculations of relative free energies of binding, one of these subsites, a tyrosine-surrounded pocket, is revealed as the preferred point of complexation. For this subsite, results consistent with experimental observations are obtained; DNP-glycine is found to bind better than DNP-serine, and this, in turn, is found to bind better than DNP-alanine. The suggested binding mode makes it possible to explain both the moderate binding affinity and the differences in binding energy among the three ligands.