Merck
  • [About the decomposition of acetamide as taxonomic marker for some species of the genus pseudomonas (author's transl)].
[About the decomposition of acetamide as taxonomic marker for some species of the genus pseudomonas (author's transl)].
Zentralblatt fur Bakteriologie, Parasitenkunde, Infektionskrankheiten und Hygiene. Erste Abteilung Originale. Reihe A: Medizinische Mikrobiologie und Parasitologie (1975-11-01)
R H Schubert, J G Esanu, F Esanu
PMID814748
ABSTRACT
The comparative examination of the acetamide-decompostition and of the acetamide-utilisation as sole source of carbon demonstrates that various groups of the Pseudomonas species with the same kind of reaction have to be distinguished. The acetamide-decomposition-test according to BUHLMANN et al. and the methylene-blue-reduction-test show homogeneous results. As a positive result of these tests the formation of ammonia has always been shown. An acetamide decomposition of this kind has been demonstrable with P. aeruginosa, P. acidovorans and many P. cepacia strains. On liquid mineral media containing acetamide P. fluorescens, P. putida and P. pseudomallei show growth although ammonia is not produced and the test according to BUHLMANN et al. as well as the methylene-blue-reduction-test result negatively. In all tests P. alcaligenes, P. marginata, P. caryophilli and with certain exceptions P. sutzeri and P. putrefaciens react negatively.
Product Description
Millipore
Acetamide Nutrient Broth, NutriSelect® Basic, powder, selective by defined nutrients for Pseudomonas aeruginosa, pack of 500 g, suitable for microbiology
Pricing and availability is not currently available.