Merck
  • Influence of stereoisomers of 4-fluoroglutamate on rat brain glutamate decarboxylase.
Influence of stereoisomers of 4-fluoroglutamate on rat brain glutamate decarboxylase.
Journal of enzyme inhibition (2000-05-16)
J Drsata, M Netopilová, V Tolman
ABSTRACT
Inhibition of rat brain glutamate decarboxylase (GAD, EC 4.1.1.15) by individual stereoisomers of 4-fluoroglutamate (4-F-Glu) and 2-fluoro-4-aminobutyrate (2-F-GABA) was studied. All stereoisomers of 4-F-Glu inhibited decarboxylation of L-glutamate catalysed by the enzyme preparation. At 1 x 10(-2) M concentration, the most potent inhibitor of GAD was D-erythro-4-F-Glu with about 70% inhibition in the presence of 1.23 x 10(-2)M L-glutamate. The inhibition by all stereoisomers was of the competitive type. Ki values ranged from 2 x 10(-3)M for the D-erythro isomer to 1.1 x 10(-2)M for the D-threo and L-erythro isomers. The influence of all stereoisomers was reversible as shown by dialysis except for a small amount in the case of the D-erythro isomer. The inhibition was independent of external pyridoxal-5'-phosphate added. No inhibition of rat brain GAD was found with 2-fluoro-4-aminobutyrate stereoisomers.