Merck
  • Comparison of hepatic and renal drug-metabolising enzyme activities in sheep given single or two-fold challenge infections with Fasciola hepatica.
Comparison of hepatic and renal drug-metabolising enzyme activities in sheep given single or two-fold challenge infections with Fasciola hepatica.
International journal for parasitology (2000-08-06)
C Calléja, K Bigot, C Eeckhoutte, P Sibille, C Boulard, P Galtier
ABSTRACT
The activity of drug-metabolising enzymes was compared in liver and kidneys of adult sheep given single or two-fold fluke infection. Fascioliasis was induced by oral administration of 200 metacercariae of Fasciola hepatica to female sheep either 10 or 20 weeks (mono-infections) or 10 and 20 weeks (bi-infection) before killing. The parasitic pathology was ascertained at autopsy and by clinical observation of animals. In the liver of both mono- and bi-infected animals, significant decreases (P<0.05) (17-44%) were observed in the microsomal content of cytochrome P450 and in the two measured P450-dependent monooxygenase activities, benzphetamine and ethylmorphine N-demethylations. Moreover, Western blot analysis of microsomes demonstrated a decrease in the expression of cytochrome P4503A subfamily correlative with that of its presumed corresponding activity ethylmorphine N-demethylase. By contrast, the conjugation of chloro-dinitrobenzene to glutathione remained unchanged in liver cytosolic fractions prepared from all these animals. In kidneys, a significant decrease (P<0.05) (30%) in microsomal cytochrome P450 level of 10-week mono-infected sheep was observed whereas there was no change in the other groups of animals. The inflammatory origin and the consequences in terms of pathology and animal productivity of the fascioliasis-induced decreases in tissue-oxidative drug metabolism are discussed, particularly in the case of adult sheep suffering repetitive infections.
Product Description
Supelco
Ethylmorphine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®