Merck
  • Quantitative liquid chromatography/mass spectrometry/mass spectrometry warfarin assay for in vitro cytochrome P450 studies.
Quantitative liquid chromatography/mass spectrometry/mass spectrometry warfarin assay for in vitro cytochrome P450 studies.
Analytical biochemistry (2001-10-25)
Z Y Zhang, B M King, Y N Wong
ABSTRACT
A sensitive assay using high-performance liquid chromatography tandem mass spectrometry (MS/MS) has been established for the quantitative analysis of cytochrome P450 form-specific activities using warfarin as a probe substrate. Four metabolites, 6-, 7-, 8-, and 10-hydroxywarfarin, were chromatographically resolved within 10 min using gradient mobile phases. The mass spectrometry was operated under negative ionization mode. The MS/MS product ion spectra of warfarin and the metabolites were generated using collision-activated dissociation and interpreted. The abundant product ions of the metabolites were selected for quantification applying multiple reaction monitoring. Quantification was based on a quadratic or power curve of the peak area ratio of the metabolite over the internal standard against the respective concentration of the metabolite. This assay has been validated from 2 to 1000 nM for 10-hydroxywarfarin and from 2 to 5000 nM for 6-, 7-, and 8-hydroxywarfarin and successfully applied to evaluate cytochrome P450-mediated drug-drug interactions in vitro using human hepatocytes and liver microsomal preparations.