Merck
  • Enantioseparation of glutethimide and its 5-OH-metabolite in capillary electrophoresis and study of selector-selectand interactions using one-dimensional rotating frame nuclear Overhauser and exchange spectroscopy.
Enantioseparation of glutethimide and its 5-OH-metabolite in capillary electrophoresis and study of selector-selectand interactions using one-dimensional rotating frame nuclear Overhauser and exchange spectroscopy.
Electrophoresis (2002-07-13)
Bezhan Chankvetadze, Naira Burjanadze, Klaus Bergander, Gottfried Blaschke
ABSTRACT
Enantioseparation of glutethimide (GT) and its 5-hydroxy metabolite (5-OH-GT) has been studied with several charged cyclodextrin (CD) derivatives. The emphasis was made on the enantiomer migration order of GT and simultaneous enantioseparation of GT and 5-OH-GT. The possible structural differences of GT complexes with three different single isomer charged CD derivatives were studied using one-dimensional rotating frame nuclear Overhauser and exchange spectroscopy (1-D ROESY).