Merck
  • Effects of inspired oxygen concentration on in vivo redox reaction of nitroxide radicals in whole mice.
Effects of inspired oxygen concentration on in vivo redox reaction of nitroxide radicals in whole mice.
Biochemical and biophysical research communications (1992-02-14)
Y Miura, H Utsumi, A Hamada
ABSTRACT
Effects of inspired oxygen concentration on metabolism of nitroxide radical were measured in whole mice by using in vivo ESR. Oxygen concentration influenced spin-clearance of nitroxide differently in abdomen and head, suggesting that either O2 concentrations or metabolic mechanisms of nitroxide in the two domains were different.