Merck
  • Evidence for lithium-sensitive inositol 4,5-bisphosphate accumulation in muscarinic cholinoceptor-stimulated cerebral-cortex slices.
Evidence for lithium-sensitive inositol 4,5-bisphosphate accumulation in muscarinic cholinoceptor-stimulated cerebral-cortex slices.
The Biochemical journal (1992-10-15)
S Jenkinson, R A Challiss, S R Nahorski
ABSTRACT
Stimulation of [3H]inositol-prelabelled rat cerebral-cortex slices with carbachol results in the accumulation of four [3H]inositol bisphosphate isomeric species, Ins(1,3)P2, Ins(1,4)P2, Ins(3,4)P2 and Ins(4,5)P2. Although the last isomer ran as a minor peak on h.p.l.c., its accumulation was dramatically enhanced in the presence of Li+ (1 mM), such that at 30 min it represented almost 35% of the total bisphosphate fraction. The accumulation of Ins(4,5)P2 appeared to be very sensitive to Li+ (EC50 = 94 +/- 3 microM), strongly implicating a Li(+)-sensitive metabolism. Evidence for this is provided from the rapid but Li(+)-sensitive decay of Ins(4,5)P2 when muscarinic-receptor stimulation is antagonized by atropine at a time when accumulations have reached a new steady state. Manipulation of phospholipase D by activators and inhibitors of protein kinase C did not suggest a role for phospholipase D hydrolysis of PtdInsP2 in the formation of Ins(4,5)P2. Attempts to reveal Ins(4,5)P2 metabolism, or indeed its synthesis from Ins(1,4,5)P3, were not successful with broken cell preparations and strongly suggest discrete compartmentation of inositol phosphate metabolism in the intact cell.