- Implication of corticosteroid receptors in the regulation of hippocampal structural plasticity.
Implication of corticosteroid receptors in the regulation of hippocampal structural plasticity.
The dentate gyrus is one of the few areas of the adult brain that continues to produce neurons and to express the embryonic polysialylated isoforms of neuronal cell adhesion molecules (PSA-NCAM). The stress hormone corticosterone exerts a complex modulation on neurogenesis and PSA-NCAM, and previous studies have shown that mature granule cells require corticosterone for their survival. Thus, the aim of our work was to investigate the respective role of the different corticosteroid receptors on these three parameters in adrenalectomized rats. It was found that treatment with a low dose of the mineralocorticoid receptor agonist, aldosterone, prevents only the adrenalectomy-induced increase in cell death. Treatment with a higher dose of aldosterone normalized cell proliferation whereas PSA-NCAM expression was normalized only by treatment with the glucocorticoid receptor agonist, RU 28362. It is concluded that stimulation of the mineralocorticoid receptor is sufficient to mediate the effects of corticosterone on neurogenesis and to protect mature cells from cell death whereas stimulation of the glucocorticoid receptor is necessary to modulate PSA-NCAM expression.