Merck
  • Host plant volatiles synergize response to sex pheromone in codling moth, Cydia pomonella.
Host plant volatiles synergize response to sex pheromone in codling moth, Cydia pomonella.
Journal of chemical ecology (2004-05-14)
Zhihua Yang, Marie Bengtsson, Peter Witzgall
ABSTRACT
Plant volatile compounds synergize attraction of codling moth males Cydia pomonella to sex pheromone (E,E)-8,10-dodecadien-1-ol (codlemone). Several apple volatiles, known to elicit a strong antennal response, were tested in a wind tunnel. Two-component blends of 1 pg/min codlemone and 100 pg/min of either racemic linalool, (E)-beta-farnesene, or (Z)-3-hexen-1-ol attracted significantly more males to the source than codlemone alone (60, 58, 56, and 37%, respectively). In comparison, a blend of codlemone and a known pheromone synergist, dodecanol, attracted 56% of the males tested. Blends of pheromone and plant volatiles in a 1:100 ratio attracted more males than 1:1 or 1:10,000 blends. Adding two or four of the most active plant compounds to codlemone did not enhance attraction over blends of codlemone plus single-plant compounds. Of the test compounds, only farnesol was attractive by itself; at a release rate of 10,000 pg/min, 16% of the males arrived at the source. However, attraction to a 1:10,000 blend of codlemone and farnesol (42%) was not significantly different from attraction to codlemone alone (37%). In contrast, a codlemone mimic, (E)-10-dodecadien-1-ol, which attracted 2% males by itself, had a strong antagonistic effect when blended in a 1:10,000 ratio with codlemone.