Merck
Chlorinated herbicide (triallate) dehalogenation by iron powder.
Chemosphere (2004-10-19)
Angela Volpe, Antonio Lopez, Giuseppe Mascolo, Antonia Detomaso
ABSTRACT
The reductive degradation of a chlorinated herbicide by iron powder was investigated at lab scale. The studied substrate was triallate (S-2,3,3-trichloroallyl di-isopropyl thiocarbamate) which contains a trichloroethylene moiety potentially reducible by zero-valent iron. Degradation reactions were carried out in batch, at 25 degrees C, in the absence of oxygen, by contacting electrolytic iron powder (size range: 20-50 microm) with a triallate aqueous solution (2.5 mgl(-1)). Herbicide decay, corresponding evolutions of TOC, TOX and chloride ion release were regularly monitored throughout the reactions. Furthermore, the main degradation by-products were identified by HPLC/MS. The results showed that, after 5 days, herbicide degradation extent was about 97% and that the reaction proceeded through the formation of a dechlorinated alkyne by-product (S-2-propinyl di-isopropyl thiocarbamate) resulting from the complete dechlorination of triallate. The subsequent reduction of such an alkyne intermediate gave S-allyl di-isopropyl thiocarbamate as main end by-product. The identified by-products suggested that dechlorination took place mainly via reductive beta-elimination. However, as traces of dichloroallyl di-isopropyl thiocarbamate were also detected, a role, although minor, was assigned even to hydrogenolysis in the overall dechlorination process.
Product Description
Supelco
Triallat, PESTANAL®, analytical standard