Merck
  • The secondary electron acceptor of photosystem I in Gloeobacter violaceus PCC 7421 is menaquinone-4 that is synthesized by a unique but unknown pathway.
The secondary electron acceptor of photosystem I in Gloeobacter violaceus PCC 7421 is menaquinone-4 that is synthesized by a unique but unknown pathway.
FEBS letters (2005-06-16)
Mamoru Mimuro, Tohru Tsuchiya, Hidetoshi Inoue, Yumiko Sakuragi, Yuka Itoh, Takanori Gotoh, Hideaki Miyashita, Donald A Bryant, Masami Kobayashi
ABSTRACT
The secondary electron acceptor of photosystem (PS) I in the cyanobacterium Gloeobacter violaceus PCC 7421 was identified as menaquinone-4 (MQ-4) by comparing high performance liquid chromatograms and absorption spectra with an authentic compound. The MQ-4 content was estimated to be two molecules per one molecule of chlorophyll (Chl) a', a constituent of P700. Comparative genomic analyses showed that six of eight men genes, encoding phylloquinone/MQ biosynthetic enzymes, are missing from the G. violaceus genome. Since G. violaceus clearly synthesizes MQ-4, the combined results indicate that this cyanobacterium must have a novel pathway for the synthesis of 1,4-dihydroxy-2-naphthoic acid.
Sigma-Aldrich
1,4-Dihydroxy-2-naphthoic acid, 97%