Merck
  • Comparative activity of three repellents against the ticks Rhipicephalus sanguineus and Argas persicus.
Comparative activity of three repellents against the ticks Rhipicephalus sanguineus and Argas persicus.
Medical and veterinary entomology (1992-01-01)
S Kumar, S Prakash, M P Kaushik, K M Rao
ABSTRACT
The residual repellent activity of N,N-diethylphenyl-acetamide (DEPA), N,N-diethyl-3-methyl benzamide (DEET) and dimethylphthalate (DMP) against the hard tick Rhipicephalus sanguineus (Latreille) and the soft tick Argas persicus (Oken) from topical application on rabbits and hens respectively in the laboratory have been studied. It was found that a 25% concentration of DEET gave strong repellency (greater than 90%) against larvae and nymphs of R. sanguineus for 15 and 7 days respectively and against A. persicus larvae and nymphs greater than 90% for 11 and 5 days respectively. Strong repellency (greater than 90%) against adults of the hard and soft ticks was observed for 9 and 7 days respectively with 40% DEET. DEPA at 25% concentration showed strong repellency (greater than 90%) against larvae and nymphs of R. sanguineus and A. persicus for 11 and 5 days respectively. Adults of hard and soft ticks were strongly repelled (greater than 90%) for 9 and 5 days respectively by 40% DEPA. DMP was found to be the least effective against all stages of both species of tick.