Merck
Synthesis of linear, branched, and cyclic peptide chimera.
Methods in molecular biology (Clifton, N.J.) (2005-07-27)
Gábor Mezö, Ferenc Hudecz
ABSTRACT
Chimeric peptides are unnatural constructs consisting of bioactive compounds from at least two different peptide(s) and/or protein(s) or two sequences from different parts of the same protein. Such multifunctional peptide combinations are prepared to enhance the biological activity or selectivity of their components. New biological effects can also be achieved with the chimera. In this chapter the synthesis of three different types of chimeric peptides will be described. In a linear chimera, two peptide epitopes from different parts of glycoprotein D (gD) of herpes simplex virus (HSV) are combined. A branched chimera, built from linear peptides, consists of tuftsin oligomers with immunostimulatory activity and an epitope peptide of HSV gD. The third compound is a cyclic chimeric molecule, where alpha-conotoxin GI as a host peptide is modified by the incorporation of a core epitope from HSV gD as a guest sequence.