Merck
  • Effects of teflubenzuron on sediment processing by members of the Capitella species-complex.
Effects of teflubenzuron on sediment processing by members of the Capitella species-complex.
Environmental pollution (Barking, Essex : 1987) (2005-08-09)
Nuria Méndez
ABSTRACT
The cosmopolitan deposit-feeding polychaete Capitella is widely used as an indicator of organic pollution and plays an important role in waste recycling at aquaculture sites. Teflubenzuron is currently employed by salmon farms as an in-feed compound to control ectoparasite infestations. Its occurrence in sediments could pose a hazard to local sediment infauna. A bioassay to investigate the effects of exposing Capitella sp I and Capitella sp B to sediment spiked with environmentally relevant concentrations of teflubenzuron (8.4, 25 and 41.8 microg/g dry wt. sediment) is described. No mortality was recorded for Capitella sp I, but increasing teflubenzuron concentrations significantly reduced egestion rates. Capitella sp B exhibited mortality, although egestion was not affected. These results using Capitella spp. indicated that teflubenzuron could affect the rate of sediment processing by polychaetes close to fish farms treated with this compound. Further studies are recommended to assess the potential of contaminants in aquaculture wastes.
Product Description
Supelco
Teflubenzuron, PESTANAL®, analytical standard
Pricing and availability is not currently available.