Merck
  • EPR study of a copper impurity center in a single crystal of 2-thiothymine.
EPR study of a copper impurity center in a single crystal of 2-thiothymine.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2005-09-17)
Erim Besić, Vjeran Gomzi, Kresimir Sanković, Janko N Herak, Dubravka Krilov
ABSTRACT
EPR spectroscopy was used to study the complex formed in crystals of 5-methyl-2-thiouracil (2-thiothymine) containing traces of copper. The copper impurities, originally present as Cu(I)-complex of 2-thiothymine in the lattice of 2-thiothymine, are transformed into paramagnetic Cu(II)-complex by ionizing radiation. It was found that the complex is planar, the plane being defined by two pairs of S and N atoms, from two adjacent 2-thiothymine molecules. The structure of the complex suggests that a pair of hydrogen bonds between two neighboring molecules is replaced by a stronger Cu-coordination bonding, with two sulfur and two nitrogen atoms as ligands. The spectroscopic parameters (g-tensor, A(63Cu) and A(14N) tensors) are essentially similar to those earlier observed for copper planar centers in other systems.