Merck
  • Applications of derivative UV spectrophotometry for the determinations of fenbufen and ketoprofen in pharmaceutical preparations.
Applications of derivative UV spectrophotometry for the determinations of fenbufen and ketoprofen in pharmaceutical preparations.
Acta poloniae pharmaceutica (2005-10-01)
Arkadiusz Pomykalski, Hanna Hopkała
ABSTRACT
A methods for the determination of fenbufen and ketoprofen in pharmaceuticals by classical spectrophotometry--zero order derivative, first and second order derivatives spectrophotometry is described. using "peak-peak" (P-P) and, "peak-zero" (P-O) measurements. The calibration curves are linear within the concentration range of 2.0 - 12.0 microg x mL(-1) for fenbufen and ketoprofen. The procedure is simple, rapid and the results are reliable.