Merck
  • Persistent organochlorines in raccoon dogs (Nyctereutes procyonoides) from Japan: Hepatic sequestration of oxychlordane.
Persistent organochlorines in raccoon dogs (Nyctereutes procyonoides) from Japan: Hepatic sequestration of oxychlordane.
Chemosphere (2006-07-15)
Tatsuya Kunisue, Nozomi Takayanagi, Toshio Tsubota, Shinsuke Tanabe
ABSTRACT
The present study determined the accumulation features of persistent organochlorines (OCs) such as polychlorinated biphenyls (PCBs), dichlorodiphenyltrichloroethane and its metabolites (DDTs), hexachlorocyclohexane isomers (HCHs), hexachlorobenzene (HCB), and chlordane compounds (CHLs) in wild raccoon dogs (RDs; Nyctereutes procyonoides) collected from Kanagawa prefecture in Japan during 2001. In livers of RDs, CHLs were remarkably dominant (20 times higher than PCBs) followed by PCBs>DDTs>HCHs>HCB, whereas the chemicals in muscles were in the order of CHLs>PCBs>HCHs>DDTs>HCB. The accumulation pattern of OCs in RDs was different from those in Japanese humans and avian species reported previously, which generally accumulate higher levels of DDTs and PCBs than CHLs. This result indicates that RDs have been exposed to relatively high levels of CHLs and have high metabolic and elimination capacity for DDTs. In fact, CHL levels in RDs were higher than those in humans and some avian species, while DDT levels in RDs were much lower than other animals. In particular, extremely high accumulation levels of oxychlordane, which is a metabolite from chlordanes and nonachlors, were observed in RD livers. The higher toxic potency of oxychlordane than parent compounds may suggest that RDs are at high risk by this metabolite. On lipid weight basis, PCBs, HCHs and HCB levels were almost similar in livers and muscles, suggesting that the tissue distribution of these compounds principally followed the lipid-dependent accumulation. However, accumulation levels of oxychlordane and p,p'-DDD in livers were significantly higher than those in muscles, and concentration ratios of liver to muscle (L/M ratios) of these compounds were greater than 1.0 in all the specimens. This phenomenon was similar to PCDD/DF congener accumulation patterns observed previously in RDs. When relationships between hepatic TEQs and L/M ratios were examined for oxychlordane and p,p'-DDD, L/M ratios for these compounds significantly increased with hepatic TEQ levels, suggesting their hepatic sequestration in TEQs-dependent manner.