Merck
  • Ubiquinol and the papaverine derivative caroverine prevent the expression of tumour- promoting factors in adenoma and carcinoma colon cancer cells induced by dietary fat.
Ubiquinol and the papaverine derivative caroverine prevent the expression of tumour- promoting factors in adenoma and carcinoma colon cancer cells induced by dietary fat.
BioFactors (Oxford, England) (2006-07-29)
Hans Nohl, Natalia Rohr-Udilova, Lars Gille, Werner Bieberschulte, Daniela Jurek, Brigitte Marian, Rolf Schulte-Hermann
ABSTRACT
High consumption of dietary fat promotes colon carcinogenesis. While this effect is well known the underlying mechanism is not understood. Fatty acid hydroperoxides (LOOH) arise from unsaturated fatty acids in the presence of oxygen and elevated temperature during food processing. An approach was made starting from the assumption that LOOH are present in dietary fats as a result of boiling. LOOH undergoes homolytic cleavage in the presence of iron. We studied their effects on gene expression in colorectal tumour cells using linoleic acid hydroperoxide (LOOH) as model compound. Addition to the medium of LT97 adenoma and SW480 carcinoma cells enhanced the production of hydrogen peroxide. Both cell lines were observed to increase VEGF and COX-II expression based on mRNA. Expression of VEGF was inhibited by caroverine and ubiquinon.