Merck
  • A randomized study comparing filgrastim versus lenograstim versus molgramostim plus chemotherapy for peripheral blood progenitor cell mobilization.
A randomized study comparing filgrastim versus lenograstim versus molgramostim plus chemotherapy for peripheral blood progenitor cell mobilization.
Bone marrow transplantation (2006-09-05)
B Kopf, U De Giorgi, B Vertogen, G Monti, A Molinari, D Turci, C Dazzi, M Leoni, A Tienghi, A Cariello, M Argnani, L Frassineti, E Scarpi, G Rosti, M Marangolo
ABSTRACT
We conducted a prospective randomized clinical trial to assess the mobilizing efficacy of filgrastim, lenograstim and molgramostim following a disease-specific chemotherapy regimen. Mobilization consisted of high-dose cyclophosphamide in 45 cases (44%), and cisplatin/ifosfamide/etoposide or vinblastine in 22 (21%), followed by randomization to either filgrastim or lenograstim or molgramostim at 5 microg/kg/day. One hundred and three patients were randomized, and 82 (79%) performed apheresis. Forty-four (43%) patients were chemonaive, whereas 59 (57%) were pretreated. A median number of one apheresis per patient (range, 1-3) was performed. The median number of CD34+ cells obtained after mobilization was 8.4 x 10(6)/kg in the filgrastim arm versus 5.8 x 10(6)/kg in the lenograstim arm versus 4.0 x 10(6)/kg in the molgramostim arm (P=0.1). A statistically significant difference was observed for the median number of days of growth factor administration in favor of lenograstim (12 days) versus filgrastim (13 days) and molgramostim (14 days) (P<0.0001) and for the subgroup of chemonaive patients (12 days) versus pretreated patients (14 days) (P<0.001). In conclusion, all three growth factors were efficacious in mobilizing peripheral blood progenitor cells with no statistically significant difference between CD34+ cell yield and the different regimens, and the time to apheresis is likely confounded by the different mobilization regimens.