Merck
Antiflammin-2 activates the human formyl-peptide receptor like 1.
TheScientificWorldJournal (2006-10-31)
Ahmad M Kamal, Richard P G Hayhoe, Anbalakan Paramasivam, Dianne Cooper, Roderick J Flower, Egle Solito, Mauro Perretti
ABSTRACT
The anti-inflammatory actions of the nonapeptide antiflammin-2, identified by homology with uteroglobin and annexin-A1 sequences, have been described in some detail, yet its mechanisms of action remain elusive. Since recent data indicate an involvement of the formyl peptide receptor (FPR)-like 1 (or FPRL-1) in the effects of annexin-A1, we have tested here the effect of antiflammin-2 with respect to this receptor family. Using HEK-293 cells expressing either human FPR and FPRL-1, and an annexin-A1 peptide as tracer ([125I-Tyr]-Ac2-26), we found that antiflammin-2 competed for binding only at FPRL-1, and not FPR, with an approximate EC50 of 1 mM. In line with data produced for the full-length protein, genuine receptor activation by antiflammin-2 was confirmed by rapid phosphorylation of extracellular-regulated kinase 1 and 2. Finally, study of the neutrophil interaction with activated endothelium under flow demonstrated an inhibitory effect of antiflammin-2, thus providing functional support to a role for the antiflammin-2/FPRL-1 anti-inflammatory axis.