Merck
  • Efficient solid-phase synthesis of a library of imidazo[1,2-a]pyridine-8-carboxamides.
Efficient solid-phase synthesis of a library of imidazo[1,2-a]pyridine-8-carboxamides.
Journal of combinatorial chemistry (2007-02-27)
Ahmed Kamal, V Devaiah, K Laxma Reddy, Rajendar, Rajesh V C R N C Shetti, N Shankaraiah
ABSTRACT
A versatile method for the solid-phase synthesis of imidazo[1,2-a]pyridine-based derivatives, imidazo[1,2-a]pyridine-8-carboxamides, has been developed. They were obtained by treatment of the amino group of the polymer-bound 2-aminonicotinate with different alpha-haloketones, followed by halogenation at the 3-position of the polymer-bound imidazo[1,2-a]pyridine. The derived polymer-bound imidazo[1,2-a]pyridines 5, 6, and 7 were finally cleaved from the solid-support with an excess of primary or secondary amines. The final crude products were purified from excess amines by solid-supported liquid-liquid extraction (SLE).
Sigma-Aldrich
Imidazo[1,2-a]pyridine, 99%