Merck
  • Supplementation of adjuvants for increasing the nutritive value and cell viability of probiotic fermented milk beverage.
Supplementation of adjuvants for increasing the nutritive value and cell viability of probiotic fermented milk beverage.
International journal of food sciences and nutrition (2009-02-26)
P Shobharani, Renu Agrawal
ABSTRACT
Probiotic are microorganisms that, upon ingestion in adequate amounts, exert a beneficial effect on the host. In the present work, the potent probiotic Leuconostoc mesenteroides was used as a starter culture in the preparation of fermented milk beverage. The product was analyzed for protein, titrable acidity, fat, total sugar, fatty acids and minerals. The viability of culture and nutrition in the product was further enhanced with supplementation of adjuvants like tryptone, casein hydrolysate, cysteine hydrochloride and ascorbic acid. After 5 days, maximum viability was observed on supplementation of tryptone (100 mg/l). The protein content was enhanced by 1.1-fold in the presence of tryptone (100 mg/l) as compared with control after 5 days of storage. Fermented milk supplemented with tryptone (100 mg/l) showed maximum bioavailability of the minerals like iron (92.05%), zinc (95.02%) and magnesium (92.04%) as compared with control. The increase in the composition of beneficial fatty acids on supplementation of adjuvants supports the therapeutic value of the product.
Product Description
Millipore
Peptone from milk solids, Refined hydrolysate
Millipore
Peptone from casein and other animal proteins, suitable for microbiology
Millipore
Proteose Peptone, Enzymatic hydrolysate
Millipore
Peptone from animal tissue, from meat
Sigma-Aldrich
Peptone from animal tissue, from meat, BioReagent, suitable for cell culture, suitable for plant cell culture
Millipore
Peptone Primatone® RL, suitable for microbiology