Merck
  • Effects of equine chorionic gonadotropin and type of ovulatory stimulus in a timed-AI protocol on reproductive responses in dairy cows.
Effects of equine chorionic gonadotropin and type of ovulatory stimulus in a timed-AI protocol on reproductive responses in dairy cows.
Theriogenology (2009-03-10)
A H Souza, S Viechnieski, F A Lima, F F Silva, R Araújo, G A Bó, M C Wiltbank, P S Baruselli
ABSTRACT
The objectives were to evaluate the effects of equine chorionic gonadotropin (eCG) supplementation (with or without eCG) and type of ovulatory stimulus (GnRH or ECP) on ovarian follicular dynamics, luteal function, and pregnancies per AI (P/AI) in Holstein cows receiving timed artificial insemination (TAI). On Day 0, 742 cows in a total of 782 breedings, received 2mg of estradiol benzoate (EB) and one intravaginal progesterone (P4) insert (CIDR). On Day 8, the CIDR was removed, and all cows were given PGF2 alpha and assigned to one of four treatments in a 2 x 2 factorial arrangement: (1) CG: GnRH 48 h later; (2) CE: ECP; (3) EG: eCG+GnRH 48 h later; (4) EE: eCG+ECP. There were significant interactions for eCG x ovulatory stimulus and eCG x BCS. Cows in the CG group were less likely (28.9% vs. 33.8%; P<0.05) to become pregnant compared with those in the EG group (odds ratio [OR]=0.28). There were no differences in P/AI between CE and EE cows (30.9% vs. 29.1%; OR=0.85; P=0.56), respectively. Thinner cows not receiving eCG had lower P/AI than thinner cows receiving eCG (15.2% vs. 38.0%; OR=0.20; P<0.01). Treatment with eCG tended to increase serum progestesterone concentrations during the diestrus following synchronized ovulation (P<0.10). However, the treatment used to induce ovulation did not affect CL volume or serum progesterone concentrations. In conclusion, both ECP and GnRH yielded comparable P/AI. However, eCG treatment at CIDR removal increased pregnancy rate in cows induced to ovulate with GnRH and in cows with lower BCS.
Sigma-Aldrich
β-Estradiol 17-cypionate