Merck
  • Oxidative and endoplasmic reticulum stress signaling are involved in dehydrocostuslactone-mediated apoptosis in human non-small cell lung cancer cells.
Oxidative and endoplasmic reticulum stress signaling are involved in dehydrocostuslactone-mediated apoptosis in human non-small cell lung cancer cells.
Lung cancer (Amsterdam, Netherlands) (2009-08-25)
Jen-Yu Hung, Ya-Ling Hsu, Wen-Chiu Ni, Ying-Ming Tsai, Chih-Jen Yang, Po-Lin Kuo, Ming-Shyan Huang
ABSTRACT
This study investigates the anticancer effect of dehydrocostuslactone (DHE), a medicinal plant-derived sesquiterpene lactone, on human non-small cell lung cancer cell lines, A549, NCI-H460 and NCI-H520. Our results show that DHE inhibits the proliferation of A549, NCI-H460 and NCI-H520 cells. DHE-induced apoptosis in both A549 and NCI-H460 cells. DHE triggered endoplasmic reticulum (ER) stress, as indicated by changes in cytosol-calcium levels, PKR-like ER kinase (PERK) phosphorylation, inositol requiring protein 1 (IRE1) and CHOP/GADD153 upregulation, X-box transcription factor-1 (XBP-1) mRNA splicing, and caspase-4 activation. The release of calcium triggered the production of ROS, which further enhances calcium overloading and subsequently activates p38, JNK and ERK1/2. Both IRE1 miRNA transfection and BAPTA-AM pretreatment inhibit DHE-mediated apoptosis, supporting the hypothesis that DHE induces cell death through ER stress. Importantly, a novel anticancer agent for the treatment of non-small cell lung cancer, and is supported by animal studies which have shown a dramatic 50% reduction in tumor size after 28 days of treatment. This study demonstrates that DHE may be a novel anticancer agent for the treatment of non-small cell lung cancer.