Merck
  • Sensitive method for detection of cocaine and associated analytes by liquid chromatography-tandem mass spectrometry in urine.
Sensitive method for detection of cocaine and associated analytes by liquid chromatography-tandem mass spectrometry in urine.
Journal of analytical toxicology (2009-10-31)
Loralie J Langman, Matthew W Bjergum, Christopher L Williamson, Frank W Crow
ABSTRACT
Cocaine (COC) is a potent CNS stimulant that is metabolized to benzoylecgonine (BE) and further metabolized to minor metabolites such as m-hydroxybenzoylecgonine (m-HOBE). COC is also metabolized to norcocaine (NC). Cocaethylene (CE) is formed when cocaine and ethyl alcohol are used simultaneously. Anhydroecgonine methyl ester (AEME) is a unique marker following smoked cocaine, and anhydroecgonine ethyl ester (AEEE) is found in cocaine smokers who also use ethyl alcohol. We developed a liquid chromatography-tandem mass spectrometry (LC-MS-MS) method for the detection and quantitation of COC, BE, NC, CE, m-HOBE, AEME, and AEEE in urine. Two hundred samples previously analyzed by gas chromatography (GC) coupled with MS were extracted using solid-phase extraction. Chromatographic separation was achieved using a gradient consisting of mobile phase A [20 mM ammonium formate (pH 2.7)] and mobile phase B (methanol/acetonitrile, 50:50), an XDB-C(8) (50 x 2.1 mm, 1.8 microm) column and a flow rate of 270 microL/min. Concentrations were calculated by comparing the peak-area with the internal standard and plotted against a standard curve. The assay displayed linearity from 1.0 to 100 ng/mL. Within- and between-run coefficients of variation were < 10% throughout the linear range. A method comparison between GC-MS and LC-MS-MS showed good correlation for COC (r(2) = 0.982) and BE (r(2) = 0.955). We report here on a sensitive method to identify clinically and forensically relevant cocaine and associated analytes at concentrations as low as 1.0 ng/mL.
Product Description
Supelco
m-Hydroxybenzoylecgonine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Anhydroecgonine methyl ester solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Norcocaine hydrochloride solution, 1.0 mg/mL in acetonitrile (as free base), ampule of 1 mL, certified reference material, Cerilliant®