Merck
  • Chromene-3-carboxamide derivatives discovered from virtual screening as potent inhibitors of the tumour maker, AKR1B10.
Chromene-3-carboxamide derivatives discovered from virtual screening as potent inhibitors of the tumour maker, AKR1B10.
Bioorganic & medicinal chemistry (2010-03-23)
Satoshi Endo, Toshiyuki Matsunaga, Kazuo Kuwata, Hai-Tao Zhao, Ossama El-Kabbani, Yukio Kitade, Akira Hara
ABSTRACT
A human aldose reductase-like protein, AKR1B10 in the aldo-keto reductase (AKR) superfamily, was recently identified as a therapeutic target in the treatment of several types of cancer. In order to identify potential leads for new inhibitors of AKR1B10, we adopted the virtual screening approach using the automated program icm, which resulted in the discovery of several chromene-3-carboxamide derivatives as potent competitive inhibitors. The most potent (Z)-2-(4-methoxyphenylimino)-7-hydroxy-N-(pyridin-2-yl)-2H-chromene-3-carboxamide inhibited the reductase activity of AKR1B10 with a K(i) value of 2.7nM, and the metabolism of farnesal and 4-hydroxynonenal in the AKR1B10-overexpressed cells from 0.1microM with an IC(50) value equal to 0.8microM.
Sigma-Aldrich
Farnesal, mixture of isomers, technical
Pricing and availability is not currently available.