Merck
  • Mechanism of action of the antiplatelet peptide, arietin, from Bitis arietans venom.
Mechanism of action of the antiplatelet peptide, arietin, from Bitis arietans venom.
Biochimica et biophysica acta (1991-05-24)
T F Huang, W J Wang, C M Teng, C Ouyang
ABSTRACT
Arietin, an Arg-Gly-Asp containing peptide from venom of Bitis arietans, inhibited aggregation of platelets stimulated by a variety of agonists with a similar IC50, 1.3-2.7.10(-7) M. It blocked aggregation through the interference of fibrinogen binding to fibrinogen receptors on platelet surface. In this paper, we further demonstrated that arietin had no significant effect on the intracellular mobilization of Ca2+ in Quin2-AM-loaded platelets stimulated by thrombin. It inhibited 125I-fibrinogen binding to ADP-stimulated platelets in a competitive manner (IC50, 1.1.10(-7) M). 125I-arietin bound to unstimulated, ADP-stimulated and elastase-treated platelets in a saturable manner and its Kd values were estimated to be 3.4.10(-7), 3.4.10(-8) and 6.5.10(-8) M, respectively, while the corresponding binding sites were 46,904, 48,958 and 34,817 per platelet, respectively. Arg-Gly-Asp-Ser (RGDS) inhibited 125I-arietin binding to ADP-stimulated platelets in a competitive manner. RGD-containing peptides, including trigramin and rhodostomin, EDTA and monoclonal antibody, 7E3, raised against glycoprotein IIb-IIIa complex, inhibited 125I-arietin binding to ADP-stimulated platelets, indicating that the binding sites of arietin appear to be located at or near glycoprotein IIb-IIIa complex. In conclusion, arietin and other RGD-containing trigramin-like peptides preferentially bind to the fibrinogen receptors associated with glycoprotein IIb-IIIa complex of the activated platelets, thus leading to the blockade of fibrinogen binding to its receptors and subsequent aggregation. The presence of RGD of arietin is essential for the expression of its biological activity. Its binding sites are overlapped with those of trigramin, rhodostomin and the monoclonal antibody, 7E3.