Merck
  • Comment on: "FTIR and FTRaman spectra, assignments, ab initio HF and DFT analysis of 4-nitrotoluene" by S. Ramalingam et al., Spectrochemica Acta A 75 (2010) 1308-1314.
Comment on: "FTIR and FTRaman spectra, assignments, ab initio HF and DFT analysis of 4-nitrotoluene" by S. Ramalingam et al., Spectrochemica Acta A 75 (2010) 1308-1314.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2010-07-06)
John Tomkinson
ABSTRACT
The title paper reports the ab initio calculated structure and vibrational spectra of 4-nitrotoluene. The calculated structure is wrong and the assignments are inconsistent with earlier work. We have recalculated the ab initio structure and reassigned the spectra in agreement with the standard text.
Sigma-Aldrich
4-Nitrotoluene, 99%