Merck
Ziconotide combination intrathecal therapy: rationale and evidence.
The Clinical journal of pain (2010-07-20)
Mark S Wallace, Richard L Rauck, Timothy Deer
ABSTRACT
Ziconotide is a nonopioid intrathecal analgesic used to manage moderate to severe chronic pain. Although ziconotide is approved in the United States for intrathecal monotherapy only, it is often used in combination with other intrathecal drugs in clinical practice. The need exists for a critical assessment of the currently available published literature on ziconotide combination therapy. This review summarizes and evaluates the publications from preclinical and clinical peer-reviewed experiments that have investigated the safety and effectiveness of ziconotide in combination with a variety of other drugs. Eleven relevant publications were identified through a systematic search of multiple databases. In preclinical studies, additive or synergistic antinociceptive effects were discovered when ziconotide was used in combination with morphine, clonidine, or baclofen; however, no additional antinociceptive effects were observed when bupivacaine was added to ziconotide therapy. Safety data from animal studies revealed that ziconotide did not exacerbate morphine-induced respiratory depression, or clonidine-induced hypotension or bradycardia; however, ziconotide did potentiate morphine-induced hypotension and inhibition of gastrointestinal tract motility. Results from 2 open-label trials indicated that combination ziconotide and morphine therapy produced greater analgesia than was produced by the use of either drug alone. Preliminary support for the use of ziconotide in combination with morphine, baclofen, or hydromorphone was provided by case studies. Although clinical and preclinical studies provide some support for the use of ziconotide in combination with morphine, hydromorphone, clonidine, or baclofen, strong evidence-based data are limited. Controlled, long-term clinical trials are warranted.