Merck
  • Chemical composition of the essential oil of Croton gossypiifolius from Venezuela.
Chemical composition of the essential oil of Croton gossypiifolius from Venezuela.
Natural product communications (2011-03-04)
Alírica I Suárez, Marly Oropeza, Luis Vásquez, Stephen Tillett, Reinaldo S Compagnone
ABSTRACT
The essential oil from leaves of Croton gossypiifolius Vahl. (Euphorbiaceae) was obtained by hydrodistillation, and analyzed by GC/FID and GC/MS. The constituents were identified by their mass spectra and Kovats' indices. Fifty-one compounds accounting for 92% of the oil were detected, and 47 of them were identified. The oil was dominated by oxygenated sesquiterpenes with the major presence of alpha-cedrene oxide (18.6%), spathulenol (16.3%), valencene (5.8%), geranyl-pentanoate (5.3%), alpha-cadinol (4.0%), germacrene D (3.5%) and longifolene (3.3%).