Merck
  • Effects of buflomedil on skin blood flow in patients with type 2 diabetes mellitus without overt micro- or macroangiopathy.
Effects of buflomedil on skin blood flow in patients with type 2 diabetes mellitus without overt micro- or macroangiopathy.
International angiology : a journal of the International Union of Angiology (2011-03-24)
D Tsantilas, A I Hatzitolios, K Tziomalos, D Karamitsos, D Papadimitriou
ABSTRACT
The aim of this study was to assess the effects of buflomedil on the peripheral microcirculation in patients with type 2 diabetes mellitus (T2DM) without overt micro- or macroangiopathy. Twenty-three patients with T2DM were randomly assigned to receive buflomedil 600 mg/day for six months (N.=12) or no medication (N.=11). Skin blood flow in the lower limbs was assessed at baseline and after 3 and 6 months using Laser Doppler. We measured the following laser Doppler parameters: volume, flow and velocity. In patients treated with buflomedil, there was a significant increase in volume (P=0.039) and a trend for an increase in both flow and velocity (P=0.097 for both parameters). In contrast, significant decreases in volume and flow were observed in the control group (P=0.045 and P=0.027, respectively) whereas velocity did not change (P=0.150). In conclusion, buflomedil appears to have a beneficial effect on the peripheral microcirculation in patients with T2DM.