Merck
  • Impact of withdrawal of the analgesic Co-proxamol on nonfatal self-poisoning in the UK.
Impact of withdrawal of the analgesic Co-proxamol on nonfatal self-poisoning in the UK.
Crisis (2011-05-24)
Keith Hawton, Helen Bergen, Keith Waters, Elizabeth Murphy, Jayne Cooper, Navneet Kapur
ABSTRACT
In early 2005 the UK Committee on Safety of Medicines (CSM) announced gradual withdrawal of the analgesic co-proxamol because of its adverse benefit/safety ratio, especially its use for intentional and accidental fatal poisoning. Prescriptions of co-proxamol were reduced in the 3-year withdrawal phase (2005 to 2007) following the CSM announcement. To assess the impact of the CSM announcement in January 2005 to withdraw co-proxamol on nonfatal self-poisoning with co-proxamol and other analgesics. Interrupted time series analysis of general hospital presentations for nonfatal self-poisoning (five hospitals in three centers in England), comparing the 3-year withdrawal period 2005-2007 with 2000-2004. A marked reduction in the number of episodes of nonfatal self-poisoning episodes involving co-proxamol was found following the CSM announcement (an estimated 62% over the period 2005 to 2007 compared to 2000 to 2004). There was no evidence of an increase in nonfatal self-poisoning episodes involving other analgesics (co-codamol, codeine, co-dydramol, dihydrocodeine, and tramadol) in relation to the CSM announcement over the same period, nor a change in the number of all episodes of self-poisoning. Data were from three centers only. The impact of the policy appears to have reduced nonfatal self-poisoning with co-proxamol without significant substitution with other analgesics. This finding is in keeping with that for suicide.
Product Description
Supelco
(+)-Propoxyphene solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®