Merck
  • Effect of the single-drop mydriatic combination of 0.8% tropicamide with 5% phenylephrine with multiple applications of the same drop: a randomized controlled trial.
Effect of the single-drop mydriatic combination of 0.8% tropicamide with 5% phenylephrine with multiple applications of the same drop: a randomized controlled trial.
Indian journal of ophthalmology (2011-06-15)
Jitendra Jethani, Himanshu Solanki, Amod Nayak
Sigma-Aldrich
Tropicamide, solid