Merck
  • Ribose 5-phosphate glycation reduces cytochrome c respiratory activity and membrane affinity.
Ribose 5-phosphate glycation reduces cytochrome c respiratory activity and membrane affinity.
Biochemistry (2011-11-19)
Gordon J Hildick-Smith, Michael C Downey, Lisa M Gretebeck, Rebecca A Gersten, Roger K Sandwick
ABSTRACT
Spontaneous glycation of bovine heart cytochrome c (cyt c) by the sugar ribose 5-phosphate (R5P) weakens the ability of the heme protein to transfer electrons in the respiratory pathway and to bind to membranes. Trypsin fragmentation studies suggest the preferential sites of glycation include Lys72 and Lys87/88 of a cationic patch involved in the association of the protein with its respiratory chain partners and with cardiolipin-containing membranes. Reaction of bovine cyt c with R5P (50 mM) for 8 h modified the protein in a manner that weakened its ability to transfer electrons to cytochrome oxidase by 60%. An 18 h treatment with R5P decreased bovine cyt c's binding affinity with cardiolipin-containing liposomes by an estimated 8-fold. A similar weaker binding of glycated cyt c was observed with mitoplasts. The reversal of the effects of R5P on membrane binding by ATP further supports an A-site modification. A significant decrease in the rate of spin state change for ferro-cyt c, thought to be due to cardiolipin insertion disrupting the coordination of Met to heme, was found for the R5P-treated cyt c. This change occurred to a greater extent than what can be explained by the permanent attachment of the protein to the liposome. Turbidity changes resulting from the multilamellar liposome fusion that is readily promoted by cyt c binding were not seen for the R5P-glycated cyt c samples. Collectively, these results demonstrate the negative impact that R5P glycation can have on critical electron transfer and membrane association functions of cyt c.
Sigma-Aldrich
D-Ribose 5-phosphate barium salt hexahydrate, ≥99.0% (TLC)