Merck
  • Reversal of oxidant-mediated biochemical injury and prompt functional recovery after prolonged single-dose crystalloid cardioplegic arrest in the infantile piglet heart by terminal warm-blood cardioplegia supplemented with phosphodiesterase III inhibitor.
Reversal of oxidant-mediated biochemical injury and prompt functional recovery after prolonged single-dose crystalloid cardioplegic arrest in the infantile piglet heart by terminal warm-blood cardioplegia supplemented with phosphodiesterase III inhibitor.
General thoracic and cardiovascular surgery (2012-02-14)
Katsushi Kinouchi, Kiyozo Morita, Yoshihiro Ko, Ryuichi Nagahori, Gen Shinohara, Takayuki Abe, Kazuhiro Hashimoto
ABSTRACT
The benefit of terminal blood cardioplegia (TWBCP) is insufficient after prolonged ischemia associated with inevitable oxidant-mediated injury by this modality alone. We tested the effects of TWBCP supplemented with high-dose olprinone, which is a phosphodiesterase III inhibitor, a clinically available compound with the potential to reduce oxidant stress and calcium overload. We evaluated the effects with respect to avoiding oxidant-mediated myocardial reperfusion injury and prompt functional recovery after prolonged single-dose crystalloid cardioplegic arrest in a infantile piglet cardiopulmonary bypass (CPB) model. Fifteen piglets were subjected to 90 min of cardioplegic arrest on CPB, followed by 30 min of reperfusion. In group I, uncontrolled reperfusion was applied without receiving TWBCP; in group II, TWBCP was given; in group III, TWBCP was supplemented with olprinone (3 μg/ml). Myocardial performance was evaluated before and after CPB by a left ventricular (LV) function curve and pressure-volume loop analyses. Biochemical injury was determined by measurements of troponin-T and lipid peroxide (LPO) in coronary sinus blood. Group III showed significant LV performance recovery (group I, 26.5% ± 5.1%; group II, 42.9% ± 10.8%; group III, 81.9% ± 24.5%, P < 0.01 vs. groups I and II), associated with significant reduction of troponin-T and LPO at the reperfusion phase. No piglets in group III needed electrical cardioversion. We concluded that TWBCP with olprinone reduces myocardial reperfusion injury by reducing oxidant-mediated lipid peroxidation, and it accelerates prompt and persistent LV functional recovery with suppression of reperfusion arrhythmia.