Merck
  • Controlled growth of narrowly dispersed nanosize hexagonal MOF rods from Mn(III)-porphyrin and In(NO3)3 and their application in olefin oxidation.
Controlled growth of narrowly dispersed nanosize hexagonal MOF rods from Mn(III)-porphyrin and In(NO3)3 and their application in olefin oxidation.
Chemical communications (Cambridge, England) (2012-04-28)
Da Hee Lee, Sundol Kim, Min Young Hyun, Jin-Yeon Hong, Seong Huh, Cheal Kim, Suk Joong Lee
ABSTRACT
A new class of narrowly dispersed nanosize hexagonal MOF rods from Mn(III)-porphyrin and In(III) was obtained. The length of MOF rods was controlled by simple change of reaction times. Furthermore, the oxidation of styrene has been successfully demonstrated with Mn(III)-porphyrin MOF rods and their reusability has been also tested.