Merck
  • Flexible pyroelectric nanogenerators using a composite structure of lead-free KNbO(3) nanowires.
Flexible pyroelectric nanogenerators using a composite structure of lead-free KNbO(3) nanowires.
Advanced materials (Deerfield Beach, Fla.) (2012-07-28)
Ya Yang, Jong Hoon Jung, Byung Kil Yun, Fang Zhang, Ken C Pradel, Wenxi Guo, Zhong Lin Wang
ABSTRACT
Pyroelectric nanogenerators fabricated using a lead-free KNbO(3) nanowire-PDMS polymer composite are reported for the first time. The voltage/current output of the nanogenerators can be controlled by electric fields and enhanced by increasing the rate of change in temperature. The fabricated nanogenerators can be used to harvest energy from sunlight illumination and have potential applications in self-powered nanodevices and nanosystems.